1-(2-Bromoethoxy)-2-ethoxybenzene 98%
Supplier: Thermo Scientific
Warning
Synonyms: 2-(2-Ethoxyphenoxy)ethyl bromide
L20300.06EA 54.5 CHF
L20300.06 L20300.14 L20300.22
1-(2-Bromoethoxy)-2-ethoxybenzene 98%
1-(2-Bromoethoxy)-2-ethoxybenzene
Formula: C₁₀H₁₃BrO₂
Melting Pt: 41…43 °C
Storage Temperature: Ambient
MDL Number: MFCD02030483
CAS Number: 3259-03-8
EINECS: 000-000-0