2-Bromo-6-nitroanisole

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1-Bromo-2-methoxy-3-nitrobenzene

ACTIAS25472G5EA 46 CHF
ACTIAS25472G5
2-Bromo-6-nitroanisole
2-Bromo-6-nitroanisole
Formula: C₇H₆BrNO₃
MW: 232.0300 g/mol
MDL Number: MFCD09261208
CAS Number: 98775-19-0
EINECS: 225-983-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR