1,4-Dibrom-2,3-difluorbenzol
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS35250G1EA
0
CHF
ACTIAS35250G1
ACTIAS35250G5
ACTIAS35250G10
1,4-Dibrom-2,3-difluorbenzol
1,4-Dibromo-2,3-difluorobenzene
Formel:
C₆H₂Br₂F₂ MW: 271,8849 g/mol |
MDL Number:
MFCD09835192 CAS-Nummer: 156682-52-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...