2'-Hydroxy-5'-nitroacetophenone

Synonyms: 1-(2-Hydroxy-5-nitrophenyl)ethanone

ACTIAS20199G10EA 0 CHF
ACTIAS20199G50 ACTIAS20199G10
2'-Hydroxy-5'-nitroacetophenone
2'-Hydroxy-5'-nitroacetophenone
Formula: C₈H₇NO₄
MW: 181.1500 g/mol
MDL Number: MFCD00463816
CAS Number: 1450-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR