(R)-(-)-2-Aminononane ≥99%, ee 98%, ChiPros®
Supplier: Thermo Scientific
Danger
L19310.06EA 203 CHF
L19310.06 L19310.14 L19310.03
(R)-(-)-2-Aminononane ≥99%, ee 98%, ChiPros®
(R)-(-)-2-Aminononane
Formula: C₉H₂₁N
MW: 143.27
Boiling Pt: 73 °C (19 mmHg)
Density: 0.782
Flash Pt: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD03844739
CAS Number: 74069-74-2
ADR: 8,II