6-Hydroxy-m-anisidin

ACTIAS35516G1EA 0 CHF
ACTIAS35516G1 ACTIAS35516G5
6-Hydroxy-m-anisidin
6-Hydroxy-m-anisidin
Formel: C₇H₉NO₂
MW: 139,16 g/mol
MDL Number: MFCD06616911
CAS-Nummer: 20734-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR