3-Chloro-5-(trifluoromethyl)benzylamine 97%
Supplier: Thermo Scientific
Danger
H34423.06EA 681 CHF
H34423.06 H34423.03
3-Chloro-5-(trifluoromethyl)benzylamine 97%
3-Chloro-5-(trifluoromethyl)benzylamine
Formula: C₈H₇ClF₃N
Storage Temperature: Ambient
MDL Number: MFCD07786683
CAS Number: 400771-41-7
ADR: 8,III