3-Chloro-4-(trifluoromethyl)benzylamine 97%

Supplier: Thermo Scientific
Danger

H31883.03EA 195 CHF
H31883.03 H31883.06
3-Chloro-4-(trifluoromethyl)benzylamine 97%
3-Chloro-4-(trifluoromethyl)benzylamine
Formula: C₈H₇ClF₃N
Storage Temperature: Ambient
MDL Number: MFCD04972759
CAS Number: 361393-93-3
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR