2-Bromo-2',4'-difluoroacetophenone ≥97%

Danger

Synonyms: 2,4-Difluorophenacyl bromide

H56442.06EA 532 CHF
H56442.06
2-Bromo-2',4'-difluoroacetophenone ≥97%
2-Bromo-2',4'-difluoroacetophenone
Formula: C₈H₅BrF₂O
MW: 235.03
Melting Pt: 31…35 °C
Flash Pt: 51 °C (124 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00142822
CAS Number: 102429-07-2
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR