2-Bromo-2',3'-difluoroacetophenone

Supplier: Apollo Scientific

Synonyms: 2,3-Difluorophenacyl bromide

APOSPC10408-5GEA 362 CHF
APOSPC10408-5G
2-Bromo-2',3'-difluoroacetophenone
2-Bromo-2',3'-difluoroacetophenone
Formula: C₈H₅BrF₂O
MDL Number: MFCD07368687
CAS Number: 886762-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR