2-Fluor-4-(trifluormethoxy)benzylamin 97%
|
Gefahr
|
H33649.MDEA
73.4
CHF
H33649.MD
H33649.03
2-Fluor-4-(trifluormethoxy)benzylamin 97%
2-Fluor-4-(trifluormethoxy)benzylamin
Formel:
C₈H₇F₄NO MW: 209,14 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD16652498 CAS-Nummer: 1240257-11-7 UN: 3267 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...