2,3-Difluoro-4-methoxybenzylamine 97%
Supplier: Alfa Aesar
Danger
H26452.MDEA 49 CHF
H26452.MD H26452.03
2,3-Difluoro-4-methoxybenzylamine 97%
2,3-Difluoro-4-methoxybenzylamine
Formula: C₈H₉F₂NO
Melting Pt: 32…35 °C
Storage Temperature: Ambient
MDL Number: MFCD04115987
CAS Number: 886500-75-0
ADR: 8,III