4-Fluor-3-methylphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS50585G5EA
0
CHF
ACTIAS50585G5
ACTIAS50585G25
ACTIAS50585G100
4-Fluor-3-methylphenylboronsäure
4-Fluor-3-methylphenylboronsäure
Formel:
C₇H₈BFO₂ MW: 153,95 g/mol |
MDL Number:
MFCD01863527 CAS-Nummer: 139911-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...