4-Fluoro-2-methylphenylboronic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
Synonyms:
4-Fluoro-2-methylbenzeneboronic acid
ACTIAS51399G5EA
0
CHF
ACTIAS51399G5
ACTIAS51399G100
ACTIAS51399G25
ACTIAS51399G1
4-Fluoro-2-methylphenylboronic acid
4-Fluoro-2-methylphenylboronic acid
Formula:
C₇H₈BFO₂ MW: 153.95 |
MDL Number:
MFCD02093072 CAS Number: 139911-29-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...