3-Fluoro-4-methylphenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-Fluoro-4-methylbenzeneboronic acid

ACTIAS50569G5EA 0 CHF
ACTIAS50569G5 ACTIAS50569G25 ACTIAS50569G1
3-Fluoro-4-methylphenylboronic acid
3-Fluoro-4-methylphenylboronic acid
Formula: C₇H₈BFO₂
MW: 153.95 g/mol
MDL Number: MFCD02683115
CAS Number: 168267-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR