2-Brom-3-nitrophenol
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS36621G5EA
0
CHF
ACTIAS36621G5
ACTIAS36621G1
ACTIAS36621MG250
2-Brom-3-nitrophenol
2-Brom-3-nitrophenol
Formel:
C₆H₄BrNO₃ MW: 218,0100 g/mol |
MDL Number:
MFCD08704555 CAS-Nummer: 101935-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...