2-Brom-5-(trifluormethyl)anisol
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS35553G1EA
0
CHF
ACTIAS35553G1
ACTIAS35553G5
2-Brom-5-(trifluormethyl)anisol
2-Brom-5-(trifluormethyl)anisol
Formel:
C₈H₆BrF₃O MW: 255,03 g/mol |
MDL Number:
MFCD04115974 CAS-Nummer: 402-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...