3-Isopropoxyphenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-Isopropoxybenzeneboronic acid

ACTIAS32787G5EA 0 CHF
ACTIAS32787G10 ACTIAS32787G1 ACTIAS32787G5
3-Isopropoxyphenylboronic acid
3-Isopropoxyphenylboronic acid
Formula: C₉H₁₃BO₃
MW: 180.01
MDL Number: MFCD03701512
CAS Number: 216485-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR