4-tert-Butylphenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-tert-Butylbenzeneboronic acid(4-(tert-Butyl)phenyl)boronic acid

ACTIAS50055G100EA 0 CHF
ACTIAS50055G100
4-tert-Butylphenylboronic acid
4-tert-Butylphenylboronic acid
Formula: C₁₀H₁₅BO₂
MW: 178.04 g/mol
MDL Number: MFCD01009697
CAS Number: 123324-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR