3-Bromocinnamic acid überwiegend trans
|
Achtung
|
ACTIAS23858G25EA
0
CHF
ACTIAS23858G25
ACTIAS23858G100
3-Bromocinnamic acid überwiegend trans
3-Bromocinnamic acid
Formel:
C₉H₇BrO₂ MW: 227,055 g/mol |
MDL Number:
MFCD00004382 CAS-Nummer: 32862-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...