Naled, Supelco®
Supplier: Merck
M-Clarity™ quality level = MQ100
45429-250MGEA 64 CHF
45429-250MG
Naled, Supelco®
Naled
Formula: C₄H₇Br₂Cl₂O₄P
MDL Number: MFCD00053227
CAS Number: 300-76-5
EINECS: 206-098-3