2-Chloro-4-(4-fluorobenzylcarbamoyl)benzeneboronic acid, 97%

H55649.MDEA 93.8 CHF
ALFAH55649.MD ALFAH55649.03
2-Chloro-4-(4-fluorobenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BClFNO3
Formula Weight: 307.51
MDL No.: MFCD20040319
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR