3-Biphenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: [1,1'-Biphenyl]-3-ylboronic acid

ACTIAS50023G5EA 0 CHF
ACTIAS50023G5 ACTIAS50023G100
3-Biphenylboronic acid
3-Biphenylboronic acid
Formula: C₁₂H₁₁BO₂
MW: 198.03 g/mol
MDL Number: MFCD01318102
CAS Number: 5122-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR