4-Methoxy-3-(trifluoromethyl)benzylamine 97%
Supplier: Thermo Scientific
Danger
H26748.03EA 93.6 CHF
H26748.03 H26748.09
4-Methoxy-3-(trifluoromethyl)benzylamine 97%
4-Methoxy-3-(trifluoromethyl)benzylamine
Formula: C₉H₁₀F₃NO
Storage Temperature: Ambient
MDL Number: MFCD06212984
CAS Number: 743408-04-0
UN: 2735
ADR: 8,III