3-Chloro-5-fluoro-4-methoxybenzylamine 97%, JRD
Supplier: Thermo Scientific
Danger
H31630.MDEA 74.2 CHF
H31630.MD H31630.03
3-Chloro-5-fluoro-4-methoxybenzylamine 97%, JRD
3-Chloro-5-fluoro-4-methoxybenzylamine
Formula: C₈H₉ClFNO
Storage Temperature: Ambient
MDL Number: MFCD07786699
CAS Number: 887581-86-4
ADR: 8,III