4,4'-Dicyanobenzhydrol

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS35941G5EA 0 CHF
ACTIAS35941G5 ACTIAS35941G1
4,4'-Dicyanobenzhydrol
4,4'-Dicyanobenzhydrol
Formula: C₁₅H₁₀N₂O
CAS Number: 134521-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR