3-Iodbenzylamin Hydrochlorid

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS35142G1EA 0 CHF
ACTIAS35142G1 ACTIAS35142G5
3-Iodbenzylamin Hydrochlorid
3-Iodbenzylamin Hydrochlorid
Formel: C₇H₉ClIN
MW: 269,5105 g/mol
MDL Number: MFCD00012857
CAS-Nummer: 3718-88-5
EINECS: 223-068-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR