Benzenesulphonyl fluoride ≥98%
Supplier: Thermo Scientific
|
Danger
|
L04689.03EA
38
CHF
L04689.03
Benzenesulphonyl fluoride ≥98%
Benzenesulphonyl fluoride
Formula:
C₆H₅FO₂S MW: 160.17 Boiling Pt: 203…204 °C Density: 1.334 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007412 CAS Number: 368-43-4 EINECS: 206-707-2 ADR: 8,II |