4-(Methylsulphonyl)phenol

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS34871G100EA 0 CHF
ACTIAS34871G100 ACTIAS34871G10
4-(Methylsulphonyl)phenol
4-(Methylsulphonyl)phenol
Formula: C₇H₈O₃S
MW: 172.20 g/mol
MDL Number: MFCD00047806
CAS Number: 14763-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR