3-Carboxy-4-fluorophenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-Carboxy-4-fluorophenylboronic Acid3-Carboxy-4-fluorobenzeneboronic acid

ACTIAS50120G5EA 0 CHF
ACTIAS50120G5 ACTIAS50120G1 ACTIAS50120G25
3-Carboxy-4-fluorophenylboronic acid
3-Carboxy-4-fluorophenylboronic acid
Formula: C₇H₆BFO₄
MW: 183.93 g/mol
MDL Number: MFCD03095129
CAS Number: 872460-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR