4-Fluoro-2-(methoxycarbonyl)benzeneboronic acid, 95%

H53254.MDEA 98.4 CHF
ALFAH53254.MD
4-Fluoro-2-(methoxycarbonyl)benzeneboronic acid, 95%
Purity: 95%
CAS No.: 871329-81-6
Molecular Formula: C8H8BFO4
Formula Weight: 197.96
Melting Point: 86-90°
MDL No.: MFCD07363762
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR