3-Fluoro-5-(3-methylphenylcarbamoyl)benzeneboronic acid, 97%

H54373.MDEA 311 CHF
ALFAH54373.MD ALFAH54373.03
3-Fluoro-5-(3-methylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H13BFNO3
Formula Weight: 273.07
MDL No.: MFCD20265253
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR