4-Aminophenylboronic acid hydrochloride

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: (4-Aminophenyl)boronic acid hydrochloride4-Aminobenzeneboronic acid hydrochloride

ACTIAS35791G5EA 0 CHF
ACTIAS35791G5 ACTIAS35791G1
4-Aminophenylboronic acid hydrochloride
4-Aminophenylboronic acid hydrochloride
Formula: C₆H₉BClNO₂
MW: 173.41 g/mol
MDL Number: MFCD03001333
CAS Number: 80460-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR