2-Aminophenylboronsäure Hydrochlorid
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS32407G5EA
0
CHF
ACTIAS32407G5
ACTIAS32407G1
2-Aminophenylboronsäure Hydrochlorid
2-Aminophenylboronsäure Hydrochlorid
Formel:
C₆H₉BClNO₂ MW: 173,4052 g/mol |
MDL Number:
MFCD02258096 CAS-Nummer: 863753-30-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...