2-Carboxyphenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Carboxybenzeneboronic acid2-Boronobenzoic acid

ACTIAS50083G5EA 0 CHF
ACTIAS50083G5
2-Carboxyphenylboronic acid
2-Carboxyphenylboronic acid
Formula: C₇H₇BO₄
MW: 165.94 g/mol
MDL Number: MFCD01318118
CAS Number: 149105-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR