4-(Ethoxycarbonyl)phenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-(Ethoxycarbonyl)benzeneboronic acid

ACTIAS50436G5EA 0 CHF
ACTIAS50436G1 ACTIAS50436G25 ACTIAS50436G5
4-(Ethoxycarbonyl)phenylboronic acid
4-(Ethoxycarbonyl)phenylboronic acid
Formula: C₉H₁₁BO₄
MW: 193.99
MDL Number: MFCD02179441
CAS Number: 4334-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR