3-(Ethoxycarbonyl)phenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-(Ethoxycarbonyl)benzeneboronic acid3-Ethoxycarbonylphenylboronic acid

ACTIAS32762G5EA 0 CHF
ACTIAS32762G1 ACTIAS32762G5 ACTIAS32762G10
3-(Ethoxycarbonyl)phenylboronic acid
3-(Ethoxycarbonyl)phenylboronic acid
Formula: C₉H₁₁BO₄
MW: 193.99 g/mol
MDL Number: MFCD02179444
CAS Number: 4334-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR