2-(Ethoxycarbonyl)phenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: (2-(Ethoxycarbonyl)phenyl)boronic acid2-(Ethoxycarbonyl)benzeneboronic acid

ACTIAS51327G5EA 0 CHF
ACTIAS51327G1 ACTIAS51327G25 ACTIAS51327G5
2-(Ethoxycarbonyl)phenylboronic acid
2-(Ethoxycarbonyl)phenylboronic acid
Formula: C₉H₁₁BO₄
MW: 193.99 g/mol
MDL Number: MFCD02179453
CAS Number: 380430-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR