5-(3-Hydroxyphenyl)-1H-tetrazole

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-(1H-Tetrazol-5-yl)phenol

ACTIAS23663G1EA 177 CHF
ACTIAS23663G1 ACTIAS23663G5
5-(3-Hydroxyphenyl)-1H-tetrazole
5-(3-Hydroxyphenyl)-1H-tetrazole
Formula: C₇H₆N₄O
MW: 162.16 g/mol
MDL Number: MFCD06797247
CAS Number: 96859-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR