Hexafluoroantimonic acid ∼65% in aqueous solution
Supplier: Thermo Scientific
Danger
Synonyms: Hydrogen hexafluoroantimonate(V)
33488.18EA 416 CHF
33488.18 33488.22 33488.14 33488.30
Hexafluoroantimonic acid ∼65% in aqueous solution
Hexafluoroantimonic acid
Formula: HF₆Sb
MW: 236.758 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011327
CAS Number: 16950-06-4
EINECS: 241-023-8
UN: 3264
ADR: 8,II