4'-(1-Imidazolyl)acetophenone

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS21539G25EA 0 CHF
ACTIAS21539G5 ACTIAS21539G25
4'-(1-Imidazolyl)acetophenone
4'-(1-Imidazolyl)acetophenone
Formula: C₁₁H₁₀N₂O
MW: 186.21 g/mol
MDL Number: MFCD00005282
CAS Number: 10041-06-2
EINECS: 233-123-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR