4-Ethoxy-3-(trifluoromethyl)phenylboronic acid ≥98%
|
Warning
|
Synonyms:
4-Ethoxy-3-(trifluoromethyl)benzeneboronic acid
H52524.03EA
837
CHF
H52524.03
H52524.MD
4-Ethoxy-3-(trifluoromethyl)phenylboronic acid ≥98%
4-Ethoxy-3-(trifluoromethyl)phenylboronic acid
Formula:
C₉H₁₀BF₃O₃ MW: 233.98 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07363789 CAS Number: 871329-83-8 |