5-Chlor-2-methylphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS51161G5EA
0
CHF
ACTIAS51161G5
ACTIAS51161G1
5-Chlor-2-methylphenylboronsäure
5-Chlor-2-methylphenylboronsäure
Formel:
C₇H₈BClO₂ MW: 170,40 g/mol |
MDL Number:
MFCD03411939 CAS-Nummer: 148839-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...