2-Chloro-5-methylphenylboronic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
Synonyms:
2-Chloro-5-methylbenzeneboronic acid
ACTIAS35175G1EA
0
CHF
ACTIAS35175G1
ACTIAS35175G10
ACTIAS35175G5
2-Chloro-5-methylphenylboronic acid
2-Chloro-5-methylphenylboronic acid
Formula:
C₇H₈BClO₂ MW: 170.40 |
MDL Number:
MFCD06659822 CAS Number: 193353-35-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...