Methyl-3-hydroxyhexanoate

Supplier: MATREYA

Synonyms: Methyl 3-hydroxyhexanoate

BTRN1748EA 0 CHF
BTRN1748 MAYA1748
Methyl-3-hydroxyhexanoate
Methyl 3-hydroxyhexanoate
Formula: C₇H₁₄O₃
MW: 146.1843 g/mol
MDL Number: MFCD00083583
CAS Number: 21188-58-9
EINECS: 244-261-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR