5-Brom-4-methylpyrimidin
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS30939G1EA
0
CHF
ACTIAS30939G1
ACTIAS30939G5
5-Brom-4-methylpyrimidin
5-Bromo-4-methylpyrimidine
Formel:
C₅H₅BrN₂ MW: 173,0100 g/mol |
MDL Number:
MFCD10000597 CAS-Nummer: 1439-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...