Ethyl-2-aminonicotinat
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS22642G25EA
87
CHF
ACTIAS22642G25
ACTIAS22642G5
ACTIAS22642G1
Ethyl-2-aminonicotinat
Ethyl-2-aminonicotinat
Formel:
C₈H₁₀N₂O₂ MW: 166,18 g/mol |
MDL Number:
MFCD01569452 CAS-Nummer: 13362-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...