Ethyl-2-chloroisonicotinate

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Ethyl 2-Chloropyridine-4-carboxylate

ACTIAS31844G5EA 0 CHF
ACTIAS31844G1 ACTIAS31844G10 ACTIAS31844G5
Ethyl-2-chloroisonicotinate
Ethyl 2-chloroisonicotinate
Formula: C₈H₈ClNO₂
MW: 185.6076 g/mol
MDL Number: MFCD04114259
CAS Number: 54453-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR