3-Isobutoxybenzoesäure

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS24171G5EA 458 CHF
ACTIAS24171G5
3-Isobutoxybenzoesäure
3-Isobutoxybenzoesäure
Formel: C₁₁H₁₄O₃
MDL Number: MFCD01993662
CAS-Nummer: 350997-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR