2-Chloro-4-methoxybenzonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS25311G5EA 136 CHF
ACTIAS25311G5
2-Chloro-4-methoxybenzonitrile
2-Chloro-4-methoxybenzonitrile
Formula: C₈H₆ClNO
MW: 167.5900 g/mol
MDL Number: MFCD00156133
CAS Number: 127666-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR